missing translation for 'onlineSavingsMsg'
Learn More

4-[3-(Dimethylamino)propoxy]-N-methylbenzylamine, 90%, Thermo Scientific™

Product Code. 10632665
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10632665

Brand: Thermo Scientific Maybridge CC59546CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 910037-04-6
Molecular Formula C13H22N2O
Molecular Weight (g/mol) 222.332
MDL Number MFCD09064994
InChI Key LFLFEMOINFHORX-UHFFFAOYSA-N
Synonym 4-3-dimethylamino propoxy-n-methylbenzylamine, dimethyl 3-4-methylamino methyl phenoxy propyl amine, n,n-dimethyl-3-4-methylamino methyl phenoxy propan-1-amine, 4-3-dimethylamino propoxy phenyl methyl methyl amine, benzenemethanamine, 4-3-dimethylamino propoxy-n-methyl
PubChem CID 24229610
IUPAC Name N,N-dimethyl-3-[4-(methylaminomethyl)phenoxy]propan-1-amine
SMILES CNCC1=CC=C(C=C1)OCCCN(C)C

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 222.33
Percent Purity 90%
Chemical Name or Material 4-[3-(Dimethylamino)propoxy]-N-methylbenzylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.