missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Chloro-5-trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride, 95%, Thermo Scientific™

Product Code. 15438885
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438885 250 mg 250mg
15428885 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438885 Supplier Thermo Scientific Alfa Aesar Supplier No. H34324.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338422-71-2
Molecular Formula C12H6Cl2F3NO3S
Molecular Weight (g/mol) 372.14
MDL Number MFCD00243963
InChI Key NEQIUPPXYAOTSF-UHFFFAOYSA-N
Synonym 4-3-chloro-5-trifluoromethyl-2-pyridinyl-oxy benzenesulfonoyl chloride, 4-3-chloro-5-trifluoromethyl-2-pyridinyl oxy benzenesulfonoyl chloride, 4-3-chloro-5-trifluoromethyl pyridin-2-yl oxy benzenesulfonyl chloride, 4-3-chloro-5-trifluoromethyl-2-pyridyloxy benzenesulfonyl chloride, 4-3-chloro-5-trifluoromethyl pyridin-2-yl oxy benzene-1-sulfonyl chloride, 4-3-chloro-5-trifluoromethyl-2-pyridinyl oxy benzenesulfonyl chloride, 4-3-chloro-5-trifluoromethyl pyridin-2-yl oxybenzenesulfonyl chloride, chlorotrifluoromethylpyridinyloxybenzenesulfonoylchloride, 3-chloro-2-4-chlorosulphonyl phenoxy-5-trifluoromethyl pyridine
PubChem CID 2783039
IUPAC Name 4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxybenzenesulfonyl chloride
SMILES FC(F)(F)C1=CC(Cl)=C(OC2=CC=C(C=C2)S(Cl)(=O)=O)N=C1

Specifications

Melting Point 69°C to 71°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 372.15
Percent Purity 95%
Chemical Name or Material 4-(3-Chloro-5-trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.