missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Bromothien-2-yl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10072024
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10072024 5 g Amber Glass Bottle 5g
10162754 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10072024 Supplier Thermo Scientific Maybridge Supplier No. CC67804DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 934570-51-1
Molecular Formula C11H7BrOS
Molecular Weight (g/mol) 267.14
MDL Number MFCD09879968
InChI Key BTYZFUJFFBNJTP-UHFFFAOYSA-N
Synonym 4-3-bromothien-2-yl benzaldehyde, 4-3-bromothiophen-2-yl benzaldehyde
PubChem CID 24229751
IUPAC Name 4-(3-bromothiophen-2-yl)benzaldehyde
SMILES C1=CC(=CC=C1C=O)C2=C(C=CS2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 267.15
Percent Purity 97%
Chemical Name or Material 4-(3-Bromothien-2-yl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.