missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Bromothien-2-yl)benzaldehyde, 97%, Thermo Scientific™

Catalog Number p-3586743
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
934570-51-1
C11H7BrOS
267.14
MFCD09879968
BTYZFUJFFBNJTP-UHFFFAOYSA-N
4-3-bromothien-2-yl benzaldehyde, 4-3-bromothiophen-2-yl benzaldehyde
24229751
4-(3-bromothiophen-2-yl)benzaldehyde
C1=CC(=CC=C1C=O)C2=C(C=CS2)Br
This item is not returnable. View return policy
934570-51-1
C11H7BrOS
267.14
MFCD09879968
BTYZFUJFFBNJTP-UHFFFAOYSA-N
4-3-bromothien-2-yl benzaldehyde, 4-3-bromothiophen-2-yl benzaldehyde
24229751
4-(3-bromothiophen-2-yl)benzaldehyde
C1=CC(=CC=C1C=O)C2=C(C=CS2)Br
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 267.15
Percent Purity 97%
Chemical Name or Material 4-(3-Bromothien-2-yl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'