missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Aminopropyl)-2,4-dihydro-3H-pyrazol-3-one, ≥90%, Thermo Scientific™

Product Code. 10651436
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10651436 5 g 5g
10540723 250 mg 250mg
10405082 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10651436 Supplier Thermo Scientific Maybridge Supplier No. CD02877DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7032-17-9
Molecular Formula C6H11N3O
Molecular Weight (g/mol) 141.174
MDL Number MFCD00053049
InChI Key LMLXMJPJCSUFAB-UHFFFAOYSA-N
Synonym 4-3-aminopropyl-2,4-dihydro-3h-pyrazol-3-one, 4-3-aminopropyl-1h-pyrazol-5 4h-one, 4-3-aminopropyl-2,4-dihydropyrazol-3-one, 4-3-aminopropyl-4,5-dihydro-1h-pyrazol-5-one, 3h-pyrazol-3-one,4-3-aminopropyl-2,4-dihydro, aminopropyldihydropyrazolone, 4-3-aminopropyl-2-pyrazolin-5-one, 4-3-aminopropyl-1,4-dihydropyrazol-5-one
PubChem CID 2776763
IUPAC Name 4-(3-aminopropyl)-1,4-dihydropyrazol-5-one
SMILES C1=NNC(=O)C1CCCN

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Quantity 5 g
Formula Weight 141.17
Percent Purity 90%
Chemical Name or Material 4-(3-Aminopropyl)-2, 4-dihydro-3H-pyrazol-3-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.