missing translation for 'onlineSavingsMsg'
Learn More

4-(3’,6’-Dimethyl-3’-heptyl)phenol Diethoxylate, TRC

Product Code. 30382778
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382778 10 mg
30386807 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382778 Supplier Toronto Research Chemicals Supplier No. TRCD47313510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119449-38-5
Molecular Formula C19H32O3
Molecular Weight (g/mol) 308.46
Synonym 2-[2-[4-(1-Ethyl-1,4-dimethylpentyl)phenoxy]ethoxy]ethanol
SMILES CCC(C)(CCC(C)C)c1ccc(OCCOCCO)cc1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H32O3/c1-5-19(4,11-10-16(2)3)17-6-8-18(9-7-17)22-15-14-21-13-12-20/h6-9,16,20H,5,10-15H2,1-4H3
Formula Weight 308.2351
Chemical Name or Material 4-(3',6'-Dimethyl-3'-heptyl)phenol Diethoxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.