missing translation for 'onlineSavingsMsg'
Learn More

4-(3,5-Dimethyl-1H-pyrazol-1-yl)-N-methylbenzylamine, 95%, Thermo Scientific™

Product Code. 10265504
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265504 1 g Amber Glass Bottle 1g
10623435 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10265504 Supplier Thermo Scientific Maybridge Supplier No. CC68246DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 937796-07-1
Molecular Formula C13H17N3
Molecular Weight (g/mol) 215.30
MDL Number MFCD08572134
InChI Key WVGUCDIRMDWHNO-UHFFFAOYSA-N
Synonym 4-3,5-dimethyl-1h-pyrazol-1-yl-n-methylbenzylamine, 4-3,5-dimethylpyrazol-1-yl phenyl methyl methyl amine, 1-4-3,5-dimethyl-1h-pyrazol-1-yl phenyl-n-methylmethanamine, 4-3,5-dimethylpyrazolyl phenyl methyl methylamine, 4-3,5-dimethyl-1h-pyrazol-1-yl benzyl-methylamine, 3,5-dimethyl-1-4-methylamino methyl phenyl-1h-pyrazole, 4-3,5-dimethyl-1h-pyrazol-1-yl phenyl methyl methyl amine, 1-4-3,5-dimethyl-1h-pyrazol-1-yl phenyl-n-methylmethylamine
PubChem CID 16640535
IUPAC Name 1-[4-(3,5-dimethylpyrazol-1-yl)phenyl]-N-methylmethanamine
SMILES CNCC1=CC=C(C=C1)N1N=C(C)C=C1C

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 215.3
Percent Purity 95%
Chemical Name or Material 4-(3, 5-Dimethyl-1H-pyrazol-1-yl)-N-methylbenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.