missing translation for 'onlineSavingsMsg'
Learn More

4-(3,4-Difluorophenyl)-1,3-thiazol-2-amine, 97%, Thermo Scientific™

Product Code. 10304132
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10304132 10 g Amber Glass Bottle 10g
10552622 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10304132 Supplier Thermo Scientific Maybridge Supplier No. BTB07014EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175135-32-7
Molecular Formula C9H6F2N2S
Molecular Weight (g/mol) 212.218
MDL Number MFCD00052876
InChI Key NDCSJUJQMRFHEX-UHFFFAOYSA-N
Synonym 4-3,4-difluorophenyl-1,3-thiazol-2-amine, 2-amino-4-3,4-difluorophenyl thiazole, 4-3,4-difluorophenyl thiazol-2-amine, 4-3,4-difluoro-phenyl-thiazol-2-ylamine, 2-amino-4-3,4-difluorophenyl-1,3-thiazole, 2-thiazolamine,4-3,4-difluorophenyl, 4-3,4-difluoro-phenyl-1,3-thiazol-2-amine, 4-3,4-difluorophenyl-1,3-thiazole-2-ylamine, pubchem14456, timtec-bb sbb005523
PubChem CID 706326
IUPAC Name 4-(3,4-difluorophenyl)-1,3-thiazol-2-amine
SMILES C1=CC(=C(C=C1C2=CSC(=N2)N)F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 212.22
Percent Purity 97%
Chemical Name or Material 4-(3, 4-Difluorophenyl)-1, 3-thiazol-2-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.