missing translation for 'onlineSavingsMsg'
Learn More

4-[[3-[2-(3-Chlorophenyl)ethyl]-2-pyridinyl]carbonyl]-1-piperidinecarboxylic Acid Ethyl Ester, TRC

Product Code. 30367120
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367120 5 mg 5mg
30358047 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30367120 Supplier Toronto Research Chemicals Supplier No. TRCC3781005MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107256-32-6
Molecular Formula C22 H25 Cl N2 O3
Molecular Weight (g/mol) 400.9
Synonym 4-[[3-[2-(3-chlorophenyl)ethyl]-2-pyridinyl]carbonyl]-1-Piperidinecarboxylic acid ethyl ester, Ethyl 4-[[3-[2-(3-chlorophenyl)ethyl]-2-pyridinyl]carbonyl]-1-piperidinecarboxylate
IUPAC Name ethyl 4-[3-[2-(3-chlorophenyl)ethyl]pyridine-2-carbonyl]piperidine-1-carboxylate
SMILES CCOC(=O)N1CCC(CC1)C(=O)c2ncccc2CCc3cccc(Cl)c3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H25ClN2O3/c1-2-28-22(27)25-13-10-18(11-14-25)21(26)20-17(6-4-12-24-20)9-8-16-5-3-7-19(23)15-16/h3-7,12,15,18H,2,8-11,13-14H2,1H3
Formula Weight 400.1554
Chemical Name or Material Ethyl 4-[[3-[2-(3-chlorophenyl)ethyl]-2-pyridinyl]carbonyl]-1-piperidinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.