Learn More
Memmert™ 4-20Ma Current Loop Interface Temperature 0-260°C Of One Pt100 In Chamber, For Ich110
Product Code. 16365247 Shop All Memmert Products
Change view
Click to view available options
Unit Size:
Each
Product Code. Unit Size
16365247 Each
1 options
Product Code. 16365247 Supplier Memmert™ Supplier No. ICH110V6
  • ONLINE EXCLUSIVE PRICE
    £409.00
    £389.51 / Pack of 1
    Save £19.49
    4% Off
 Restricted Item
Estimated Shipment: 19-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Memmert™ Temperature Output for external temperature monitoring.
  • 4 to 20 mA current loop interface
  • Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring
Specifications
For Use With (Equipment) Models 30, 55, 75, 110, 160, 260, 450, 750, 1060
Type Current Loop Interface
Amperage 4 to 20 mA