Learn More
Memmert™ 4-20 mA Current Loop Interface for UN110plus, Actual Value
Product Code. 17569835 Shop All Memmert Products
Click to view available options
Amperage:
4 to 20 mA
Unit Size:
Each
Product Code. 17569835
Brand: Memmert™ UN110plusV3
Only null left
Add to basket
Add to basket
Temperature Output for external temperature monitoring.
  • Temperature controller actual value
Specifications
For Use With (Equipment) UN110plus Oven
Type Current Loop Interface
Amperage 4 to 20 mA