Learn More
Memmert™ 4-20 mA Current Loop Interface for UF160plus
Shop All Memmert Products
Click to view available options
Amperage:
4 to 20 mA
Unit Size:
Each
Description
- Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring (max. 1 SingleDISPLAY, max. 3 TwinDISPLAY)
Specifications
Specifications
| For Use With (Equipment) | UF160plus Oven |
| Type | Current Loop Interface |
| Amperage | 4 to 20 mA |