missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ 4 – 20 mA Current Loop Interface CO2 Controller
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
ICO105
ICO150
ICO240
ICO50
Unit Size:
Each
Description
- Accessory
- Actual value (0 – 25 % CO2 4 – 20 mA)
Specifications
Specifications
| For Use With (Equipment) | ICO150 |
| Type | CO2 Controller |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction