missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Thienyl)benzenesulfonyl chloride, 95%, Thermo Scientific™

Product Code. 15289636 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15289636 5 g 5g
15279636 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15289636 Supplier Thermo Scientific Acros Supplier No. 451160050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 209592-91-6
Molecular Formula C10H7ClO2S2
Molecular Weight (g/mol) 258.75
InChI Key IRDZCMUQNGEQEP-UHFFFAOYSA-N
Synonym 4-2-thienyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-2-thienyl, 4-thiophen-2-yl benzene-1-sulfonyl chloride, 4-thiophen-2-yl benzenesulfonyl chloride
PubChem CID 18387759
IUPAC Name 4-thiophen-2-ylbenzenesulfonyl chloride
SMILES C1=CSC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Melting Point 114°C to 115°C
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 258.75
Percent Purity 95%
Chemical Name or Material 4-(2-Thienyl)benzenesulfonyl chloride
compliance-icons compliance-icons
Product Identifier
  • 4-(2-Thienyl)benzenesulfonyl chloride, 95%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.