missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Pyridylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15430146
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15430146 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15430146 Supplier Thermo Scientific Alfa Aesar Supplier No. H52843.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-(2-Pyridylcarbamoyl)benzeneboronic acid is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 850568-25-1
Melting Point 124°C to 128°C
Molecular Formula C12H11BN2O3
MDL Number MFCD06659876
Quantity 250 mg
Synonym 4-pyridin-2-yl aminocarbonylphenylboronic acid, 4-pyridin-2-yl-aminocarbonyl benzeneboronic acid, 4-pyridin-2-ylcarbamoyl phenyl boronic acid, 4-2-pyridylcarbamoyl benzeneboronic acid, 4-pyridin-2-yl carbamoyl phenylboronic acid, acmc-209q1n, n-2-pyridinyl 4-boronobenzamide, 4-pyridin-2-ylcarbamoyl phenylboronic acid, 4-pyridin-2-yl carbamoyl phenyl boronic acid, 4-2-pyridylaminocarbonyl phenylboronic acid
InChI Key LEAZDTWQWNXKJM-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)C(=O)NC2=CC=CC=N2)(O)O
IUPAC Name [4-(pyridin-2-ylcarbamoyl)phenyl]boronic acid
Molecular Weight (g/mol) 242.041
PubChem CID 44119539
Formula Weight 242.04
Percent Purity 97%
Chemical Name or Material 4-(2-Pyridylcarbamoyl)benzeneboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.