missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Pyridylazo)resorcinol monosodium salt monohydrate

Product Code. 11399449
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399449 1 g 1g
11429256 5 g 5g
11439256 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399449 Supplier Thermo Scientific Alfa Aesar Supplier No. A18103.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16593-81-0 | C11H9N3O2 | 215.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16593-81-0
Molecular Formula C11H9N3O2
Molecular Weight (g/mol) 215.21
MDL Number MFCD00150712,MFCD00150712,MFCD00150712,MFCD00006256
InChI Key VLCAILLZPUINNF-UKTHLTGXSA-N
Synonym 4-2-pyridylazo resorcinol monosodium saltmonohydrate
PubChem CID 132988475
IUPAC Name 3-hydroxy-4-(pyridin-2-ylhydrazinylidene)cyclohexa-2,5-dien-1-one;sodium;hydrate
SMILES OC1=CC(=O)C=C\C1=N/NC1=CC=CC=N1

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 3825637
Formula Weight 255.21 (237.19 Anhydrous)
Chemical Name or Material 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.