missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, 99+%

Product Code. 10592771 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10592771 5 g Glass bottle 5g
10144050 25 g Glass bottle 25g
10239153 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10592771 Supplier Thermo Scientific Chemicals Supplier No. 176320050
Outlet Corner
Use promo code 28080
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16593-81-0 | C11H9N3O2 | 215.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate
CAS 16593-81-0, 1141-59-9
CAS Min % 99.0
CAS Max % 100.0
Color Brown to Brown-Orange
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C11H9N3O2
MDL Number MFCD00150712,MFCD00150712,MFCD00150712,MFCD00006256
Quantity 5 g
Beilstein 22, I, 694
Merck Index 15, 7125
Synonym 4-2-pyridylazo resorcinol monosodium saltmonohydrate
Solubility Information (0.1% in water) Clear
InChI Key VLCAILLZPUINNF-UKTHLTGXSA-N
SMILES OC1=CC(=O)C=C\C1=N/NC1=CC=CC=N1
IUPAC Name 3-hydroxy-4-(pyridin-2-ylhydrazinylidene)cyclohexa-2,5-dien-1-one;sodium;hydrate
Molecular Weight (g/mol) 215.21
PubChem CID 132988475
Formula Weight 255.21
Percent Purity 99+%
Physical Form Crystalline Powder
Show More Show Less
compliance-icons
Product Identifier
  • 4-(2-Pyridylazo)resorcinol monosodium salt monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.