missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride, 97%

Product Code. 15438945
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15438945

Brand: Thermo Scientific Alfa Aesar H34441.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112539-09-0 | C10H10ClNO3S | 259.704 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112539-09-0
Molecular Formula C10H10ClNO3S
Molecular Weight (g/mol) 259.704
MDL Number MFCD00219329
InChI Key OUAVTZYGHGCCCB-UHFFFAOYSA-N
PubChem CID 2794593
IUPAC Name 4-(2-oxopyrrolidin-1-yl)benzenesulfonyl chloride
SMILES C1CC(=O)N(C1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Melting Point 104°C to 105°C
Quantity 1 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Synonym 4-2-oxopyrrolidin-1-yl benzenesulfonyl chloride, 4-2-oxo-1-pyrrolidinyl benzenesulfonyl chloride, 4-2-oxo-pyrrolidin-1-yl-benzenesulfonylchloride, 4-2-oxopyrrolidin-1-yl benzene-1-sulfonyl chloride, 4-2-oxo-pyrrolidin-1-yl-benzenesulfonyl chloride, benzenesulfonylchloride, 4-2-oxo-1-pyrrolidinyl, 1-4-chlorosulfonyl phenyl pyrrolidin-2-one, pubchem5680, acmc-1bri4, benzenesulfonylchloride,4-2-oxo-1-pyrrolidinyl
Formula Weight 259.71
Percent Purity 97%
Chemical Name or Material 4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.