missing translation for 'onlineSavingsMsg'
Learn More

[4-(2-Morpholin-4-ylethoxy)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10422833
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10422833 1 g 1g
10187602 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10422833 Supplier Thermo Scientific Maybridge Supplier No. CC42109DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 852180-76-8
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.30
MDL Number MFCD07368553
InChI Key LZRLXLXSGAEQLA-UHFFFAOYSA-N
Synonym 4-2-morpholin-4-ylethoxy phenyl methanol, 4-2-morpholin-4-yl ethoxy phenyl methanol, 4-2-morpholinoethoxy phenyl methanol, benzenemethanol,4-2-4-morpholinyl ethoxy, 4-2-morpholin-4-yl-ethoxy-phenyl-methanol, 4-2-morpholin-4-ylethoxy phenyl methan-1-ol
PubChem CID 7060580
IUPAC Name [4-(2-morpholin-4-ylethoxy)phenyl]methanol
SMILES OCC1=CC=C(OCCN2CCOCC2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 237.3
Percent Purity 97%
Chemical Name or Material [4-(2-Morpholin-4-ylethoxy)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.