missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methyl-benzoimidazol-1-yl)-butanoic acid 97+%, Thermo Scientific™

Product Code. 10460744
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10460744 10 g 10g
10093753 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10460744 Supplier Thermo Scientific Maybridge Supplier No. MO07744EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 115444-73-0
Molecular Formula C12H14N2O2
Molecular Weight (g/mol) 218.26
MDL Number MFCD00879013
InChI Key IRBRKNDBYUBKIR-UHFFFAOYSA-N
Synonym 4-2-methyl-1h-benzimidazol-1-yl butanoic acid, 4-2-methyl-benzoimidazol-1-yl-butyric acid, 1h-benzimidazole-1-butanoicacid, 2-methyl, 4-2-methylbenzimidazol-1-yl butanoic acid, 4-2-methyl-1,3-benzodiazol-1-yl butanoic acid, 4-2-methylbenzimidazolyl butanoic acid, acmc-1c9be, 1-3-carboxypropyl-2-methylbenzimidazole, 4-2-methyl-1-benzimidazolyl butanoic acid, 4-2-methyl-1h-benzo d imidazol-1-yl butanoic acid
PubChem CID 4379684
IUPAC Name 4-(2-methylbenzimidazol-1-yl)butanoic acid
SMILES CC1=NC2=CC=CC=C2N1CCCC(O)=O

Specifications

Assay Percent Range 90%
Quantity 10 g
Formula Weight 218.25
Percent Purity 90%
Chemical Name or Material 4-(2-Methyl-1H-benzimidazol-1-yl)butanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.