missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methyl-1H-imidazol-1-yl)benzonitrile, 97%, Thermo Scientific™

Product Code. 10082204
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10082204 1 g Amber Glass Bottle 1g
10595554 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10082204 Supplier Thermo Scientific Maybridge Supplier No. CC61416DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 122957-50-0
Molecular Formula C11H9N3
Molecular Weight (g/mol) 183.21
MDL Number MFCD08559033
InChI Key FAMJRKMLLLJOGZ-UHFFFAOYSA-N
PubChem CID 15005384
IUPAC Name 4-(2-methyl-1H-imidazol-1-yl)benzonitrile
SMILES CC1=NC=CN1C1=CC=C(C=C1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4-2-methyl-1h-imidazol-1-yl benzonitrile, 4-2-methylimidazol-1-yl benzonitrile, 1-4-cyanophenyl-2-methylimidazole, benzonitrile,4-2-methyl-1h-imidazol-1-yl, benzonitrile, 4-2-methyl-1h-imidazol-1-yl, 4-2-methylimidazolyl benzenecarbonitrile, acmc-1c844
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 4-(2-Methyl-1H-imidazol-1-yl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.