missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methyl-1H-imidazol-1-yl)benzoic acid hydrochloride hydrate, 95%, Thermo Scientific™

Product Code. 10285894
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10285894 1 g Amber Glass Bottle 1g
10595744 250 mg Amber Glass Bottle 250mg
10791987 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10285894 Supplier Thermo Scientific Maybridge Supplier No. CC61451DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 921938-78-5
Molecular Formula C11H13ClN2O3
Molecular Weight (g/mol) 256.686
InChI Key PSHJNJCAKINUEP-UHFFFAOYSA-N
Synonym 4-2-methyl-1h-imidazol-1-yl benzoic acid hydrochloride hydrate, 4-2-methylimidazol-1-yl benzoic acid hydrate hydrochloride
PubChem CID 44118781
IUPAC Name 4-(2-methylimidazol-1-yl)benzoic acid;hydrate;hydrochloride
SMILES CC1=NC=CN1C2=CC=C(C=C2)C(=O)O.O.Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 256.69
Percent Purity 95%
Chemical Name or Material 4-(2-Methyl-1H-imidazol-1-yl)benzoic acid hydrochloride hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.