missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methyl-1,3-thiazol-4-yl)benzoyl chloride, Tech., Thermo Scientific™

Product Code. 10709025
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 857283-93-3
Molecular Formula C11H8ClNOS
Molecular Weight (g/mol) 237.701
MDL Number MFCD08271902
InChI Key WJQGMUPBMHWAEX-UHFFFAOYSA-N
Synonym 4-2-methyl-1,3-thiazol-4-yl benzoyl chloride, benzoyl chloride,4-2-methyl-4-thiazolyl, 4-2-methylthiazol-4-yl benzoyl chloride
PubChem CID 7537526
IUPAC Name 4-(2-methyl-1,3-thiazol-4-yl)benzoyl chloride
SMILES CC1=NC(=CS1)C2=CC=C(C=C2)C(=O)Cl

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 237.71
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-(2-Methyl-1, 3-thiazol-4-yl)benzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'