missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methoxyphenyl)piperidine, 97%, Thermo Scientific™

Product Code. 10299852
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10299852 1 g Amber Glass Bottle 1g
10431683 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10299852 Supplier Thermo Scientific Maybridge Supplier No. BTB13447DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 58333-75-8
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD00277219
InChI Key SRAVSVBVHDLLPO-UHFFFAOYSA-N
PubChem CID 544738
IUPAC Name 4-(2-methoxyphenyl)piperidine
SMILES COC1=CC=CC=C1C2CCNCC2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4-2-methoxyphenyl piperidine, 4-2-methoxy-phenyl-piperidine, 4-2-methoxyphenyl-piperidine, piperidine, 4-2-methoxyphenyl, 2-methoxy-1-4-piperidyl benzene, pubchem17609, piperidine,4-2-methoxyphenyl
Formula Weight 191.27
Percent Purity 97%
Chemical Name or Material 4-(2-Methoxyphenyl)piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.