missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Keto-1-benzimidazolinyl)piperidine Hydrochloride, TRC

Product Code. 30400350
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400350 2.5 g 2.5g
30399737 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400350 Supplier Toronto Research Chemicals Supplier No. TRCK1750552.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6961-12-2
Molecular Formula C12H16ClN3O
Molecular Weight (g/mol) 253.73
Synonym 1,3-Dihydro-1-(4-piperidinyl)-2H-benzimidazol-2-one Hydrochloride, 1-(4-Piperidyl)-2-benzimidazolinone Hydrochloride, 1,3-Dihydro-1-(4-piperidinyl)benzimidazol-2-one Hydrochloride, 1-(4-Piperidinyl)-1,3-dihydro-2H-benzimidazol-2-one Hydrochloride, 4-(2-Oxo-1,3-dihydro-benzimidazol-1-yl)piperidine Hydrochloride, 4-(2-Oxo-1-benzimidazolinyl)piperidine Hydrochloride, 4-(2-Oxo-1-benzimidazoly)piperidine Hydrochloride, 4-(2-Oxo-1-benzimidazolyl)piperidine Hydrochloride
SMILES CCCCC(CC)COC(=O)c1c(Br)c(Br)c(Br)c(Br)c1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C15H18Br4O2/c1-3-5-6-9(4-2)8-21-15(20)10-7-11(16)13(18)14(19)12(10)17/h7,9H,3-6,8H2,1-2H3
Chemical Name or Material 4-(2-Keto-1-benzimidazolinyl)piperidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.