missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Iodophenoxy)tetrahydropyran, 97%, Thermo Scientific™

Product Code. 10387154
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10387154 5 g Amber Glass Bottle 5g
10776725 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10387154 Supplier Thermo Scientific Maybridge Supplier No. CC52211DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-70-3
Molecular Formula C11H13IO2
Molecular Weight (g/mol) 304.127
MDL Number MFCD09064961
InChI Key ADTBIBCNKWWYRD-UHFFFAOYSA-N
Synonym 4-2-iodophenoxy tetrahydropyran, 4-2-iodophenoxy oxane, 4-2-iodophenoxy tetrahydro-2h-pyran
PubChem CID 21981312
IUPAC Name 4-(2-iodophenoxy)oxane
SMILES C1COCCC1OC2=CC=CC=C2I

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 304.12
Percent Purity 97%
Chemical Name or Material 4-(2-Iodophenoxy)tetrahydropyran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.