missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyethyl)benzaldehyde, TRC

Product Code. 30393029
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393029 500 mg 500mg
30399260 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30393029 Supplier Toronto Research Chemicals Supplier No. TRCH827400500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163164-47-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.17
Synonym 4-(2-Hydroxyethyl)phenylcarboxaldehyde
IUPAC Name 4-(2-hydroxyethyl)benzaldehyde
SMILES OCCc1ccc(C=O)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H10O2/c10-6-5-8-1-3-9(7-11)4-2-8/h1-4,7,10H,5-6H2
Formula Weight 150.0681
Chemical Name or Material 4-(2-Hydroxyethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.