missing translation for 'onlineSavingsMsg'
Learn More

4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzaldehyde Hemifumarate, TRC

Product Code. 30393470
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393470 500 mg 500mg
30389866 250 mg 250mg
30393623 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393470 Supplier Toronto Research Chemicals Supplier No. TRCH802150500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzaldehyde Hemifumarate
Analyte or Component Names 4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzaldehyde Hemifumarate
Shipping Condition Room Temperature
Molecular Formula C26H38N2O6 • C4H4O4
InChI Formula InChI=1S/2C13H19NO3.C4H4O4/c2*1-10(2)14-7-12(16)9-17-13-5-3-11(8-15)4-6-13;5-3(6)1-2-4(7)8/h2*3-6,8,10,12,14,16H,7,9H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;;2-1+
Solution Type Neat
SMILES O=CC1=CC=C(OCC(O)CNC(C)C)C=C1.O=CC2=CC=C(OCC(O)CNC(C)C)C=C2.OC(/C=C/C(O)=O)=O
IUPAC Name 4-(2-hydroxy-3-(isopropylamino)propoxy)benzaldehyde hemifumarate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 590.67
Quantity 500 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.