missing translation for 'onlineSavingsMsg'
Learn More

[4-(2-Furyl)phenyl]methanol, ≥97%, Thermo Scientific™

Product Code. 10235504
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10235504 10 g 10g
10432643 250 mg 250mg
10451314 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10235504 Supplier Thermo Scientific Maybridge Supplier No. CC34409EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17920-85-3
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD06203036
InChI Key IKZLKNNNCKXCKP-UHFFFAOYSA-N
Synonym 4-2-furyl phenyl methanol, 4-furan-2-yl phenyl methanol, 4-fur-2-yl phenyl methanol, 4-furan-2-yl benzyl alcohol, 4-2-furyl phenyl methan-1-ol
PubChem CID 7537506
IUPAC Name [4-(furan-2-yl)phenyl]methanol
SMILES OCC1=CC=C(C=C1)C1=CC=CO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 174.2
Percent Purity 97%
Chemical Name or Material [4-(2-Furyl)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.