missing translation for 'onlineSavingsMsg'
Learn More

[4-(2-Furyl)phenyl]methanol, ≥97%, Thermo Scientific™

Catalog Number p-3175222
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
17920-85-3
C11H10O2
174.20
MFCD06203036
IKZLKNNNCKXCKP-UHFFFAOYSA-N
4-2-furyl phenyl methanol, 4-furan-2-yl phenyl methanol, 4-fur-2-yl phenyl methanol, 4-furan-2-yl benzyl alcohol, 4-2-furyl phenyl methan-1-ol
7537506
[4-(furan-2-yl)phenyl]methanol
OCC1=CC=C(C=C1)C1=CC=CO1
This item is not returnable. View return policy
17920-85-3
C11H10O2
174.20
MFCD06203036
IKZLKNNNCKXCKP-UHFFFAOYSA-N
4-2-furyl phenyl methanol, 4-furan-2-yl phenyl methanol, 4-fur-2-yl phenyl methanol, 4-furan-2-yl benzyl alcohol, 4-2-furyl phenyl methan-1-ol
7537506
[4-(furan-2-yl)phenyl]methanol
OCC1=CC=C(C=C1)C1=CC=CO1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 174.2
Percent Purity 97%
Chemical Name or Material [4-(2-Furyl)phenyl]methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'