missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Furyl)-3-buten-2-one, cis + trans, 98%

Product Code. 11484037
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484037 25 g 25g
11474037 5 g 5g
11478900 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484037 Supplier Thermo Scientific Alfa Aesar Supplier No. A19456.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-15-4 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-15-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00039566
InChI Key GBKGJMYPQZODMI-SNAWJCMRSA-N
Synonym furfurylideneacetone, 4-2-furyl-3-buten-2-one, 3-buten-2-one, 4-2-furanyl, fam polymer, oramin r, furfural acetone, oramin special gr, furfuralacetone polymer, polyfurfurylidineacetone, furfuralacetone
PubChem CID 735940
IUPAC Name (E)-4-(furan-2-yl)but-3-en-2-one
SMILES CC(=O)C=CC1=CC=CO1

Specifications

Melting Point 33°C to 38°C
Boiling Point 112°C to 115°C (10 mmHg)
Flash Point 103°C (217°F)
Quantity 25 g
Beilstein 109696
Sensitivity Light sensitive
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 4-(2-Furyl)-3-buten-2-one, cis + trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.