missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(Dimethylamino)ethoxy]benzaldehyde, ≥95%, Thermo Scientific™

Product Code. 10020274
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10020274 1 g 1g
10011634 250 mg 250mg
10356714 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10020274 Supplier Thermo Scientific Maybridge Supplier No. CC51504DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 15182-92-0
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD07368978
InChI Key CBOKAZFQZOQTOC-UHFFFAOYSA-N
Synonym 4-2-dimethylamino ethoxy benzaldehyde, p-2-dimethylamino ethoxy benzaldehyde, 4-2-dimethylamino-ethoxy-benzaldehyde, pubchem12557, 4-2-dimethylaminoethoxy benzaldehyde, 4-2-dimethylamino-ethoxy benzaldehyde, 4-2-dimethylaminoethyloxy benzaldehyde, 4-n,n-dimethylaminoethoxy benzaldehyde, 4-2-dimethylamino-1-ethoxy benzaldehyde
PubChem CID 84823
IUPAC Name 4-[2-(dimethylamino)ethoxy]benzaldehyde
SMILES CN(C)CCOC1=CC=C(C=C1)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 193.24
Percent Purity 95%
Chemical Name or Material 4-[2-(Dimethylamino)ethoxy]benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.