missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(Chloromethyl)-4-(trifluoromethyl)phenyl]morpholine, 97%, Thermo Scientific™

Product Code. 10421124
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10421124 1 g Amber Glass Bottle 1g
11356805 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10421124 Supplier Thermo Scientific Maybridge Supplier No. CC57824DA
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 892502-15-7
Molecular Formula C12H13ClF3NO
Molecular Weight (g/mol) 279.687
MDL Number MFCD09025889
InChI Key KCGVSWOLBMMCMW-UHFFFAOYSA-N
Synonym 4-2-chloromethyl-4-trifluoromethyl phenyl morpholine, morpholine,4-2-chloromethyl-4-trifluoromethyl phenyl
PubChem CID 18525887
IUPAC Name 4-[2-(chloromethyl)-4-(trifluoromethyl)phenyl]morpholine
SMILES C1COCCN1C2=C(C=C(C=C2)C(F)(F)F)CCl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 279.69
Percent Purity 97%
Chemical Name or Material 4-[2-(Chloromethyl)-4-(trifluoromethyl)phenyl]morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.