missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Chloroethyl)benzoic acid, 97%

Product Code. 11346776
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346776 10 g 10g
11356776 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346776 Supplier Thermo Scientific Alfa Aesar Supplier No. L04068.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 20849-78-9 | C9H9ClO2 | 184.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20849-78-9
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00013995
InChI Key OOAPBGPLZAFZSO-UHFFFAOYSA-N
PubChem CID 88712
IUPAC Name 4-(2-chloroethyl)benzoic acid
SMILES OC(=O)C1=CC=C(CCCl)C=C1

Specifications

Melting Point 187°C to 189°C
Quantity 10 g
Beilstein 2084041
Synonym 4-2-chloroethyl benzoic acid, benzoic acid, 4-2-chloroethyl, acmc-1cklh, 4-carboxyphenethyl chloride, p-b-chloroethyl benzoic acid, 4-2-chloro-ethyl benzoic acid, 4-2-chloro-ethyl-benzoic acid, benzoic acid,4-2-chloroethyl, 1-4-carboxyphenyl-2-chloroethane, p-beta-chloroethyl benzoic acid
Formula Weight 184.62
Percent Purity 97%
Chemical Name or Material 4-(2-Chloroethyl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.