missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Bromophenyl)-2-methyl-1,3-thiazole, 97%, Thermo Scientific™

Product Code. 10142984
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10142984 5 g Amber Glass Bottle 5g
10336284 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10142984 Supplier Thermo Scientific Maybridge Supplier No. CC68410DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 941717-01-7
Molecular Formula C10H8BrNS
Molecular Weight (g/mol) 254.15
MDL Number MFCD09879981
InChI Key SXJQVZDDCODIST-UHFFFAOYSA-N
Synonym 4-2-bromophenyl-2-methyl-1,3-thiazole, 4-2-bromophenyl-2-methylthiazole
PubChem CID 24229764
IUPAC Name 4-(2-bromophenyl)-2-methyl-1,3-thiazole
SMILES CC1=NC(=CS1)C1=CC=CC=C1Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 254.15
Percent Purity 97%
Chemical Name or Material 4-(2-Bromophenyl)-2-methyl-1, 3-thiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.