missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(Boc-amino)ethyl]aniline, 97%

Product Code. 15459968
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459968 5 g 5g
15469968 250 mg 250mg
15449968 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15459968 Supplier Thermo Scientific Alfa Aesar Supplier No. H63324.06
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 94838-59-2 | C13H20N2O2 | 236.315 g/mol

4-[2-(Boc-amino)ethyl]aniline is used as a intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-[2-(Boc-amino)ethyl]aniline is used as a intermediate for chemical research.

Solubility
Slightly soluble in water.

Notes
Refrigerate -20°C. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94838-59-2
Molecular Formula C13H20N2O2
Molecular Weight (g/mol) 236.315
MDL Number MFCD05663961
InChI Key HOPALBZGTWDOTL-UHFFFAOYSA-N
Synonym tert-butyl 4-aminophenethylcarbamate, 4-2-boc-amino ethyl aniline, 2-4-amino-phenyl-ethyl-carbamic acid tert-butyl ester, tert-butyl n-2-4-aminophenyl ethyl carbamate, 2-4-aminophenyl ethyl carbamic acid tert-butyl ester, tert-butyl 4-aminophenethyl carbamate, tert-butyl-2-4-aminophenyl ethylcarbamate, 2-methyl-2-propanyl 2-4-aminophenyl ethyl carbamate, carbamic acid, n-2-4-aminophenyl ethyl-, 1,1-dimethylethyl ester, acmc-209ru6
PubChem CID 382207
IUPAC Name tert-butyl N-[2-(4-aminophenyl)ethyl]carbamate
SMILES CC(C)(C)OC(=O)NCCC1=CC=C(C=C1)N

Specifications

Color Brown
UN Number UN2811
Solubility Information Slightly soluble in water.
Quantity 5 g
Formula Weight 236.31
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-[2-(Boc-amino)ethyl]aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.