Learn More
4-[2-(Boc-amino)ethyl]aniline, 97%
Description
4-[2-(Boc-amino)ethyl]aniline is used as a intermediate for chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-[2-(Boc-amino)ethyl]aniline is used as a intermediate for chemical research.
Solubility
Slightly soluble in water.
Notes
Refrigerate -20°C. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Specifications
| Color | Brown |
| UN Number | UN2811 |
| Solubility Information | Slightly soluble in water. |
| Quantity | 1 g |
| Formula Weight | 236.31 |
| Percent Purity | 97% |
| Physical Form | Solid |
| Chemical Name or Material | 4-[2-(Boc-amino)ethyl]aniline |
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.