missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(Boc-amino)-4-thiazolyl]benzamidoxime, 97%, Thermo Scientific™

Product Code. 15480478
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15480478

Brand: Thermo Scientific Alfa Aesar H54063.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1824739-91-4
Molecular Formula C15H18N4O3S
Molecular Weight (g/mol) 334.394
MDL Number MFCD20265169
InChI Key UERHOGGYEPCACK-UHFFFAOYSA-N
Synonym 4-2-boc-amino-4-thiazolyl benzamidoxime
PubChem CID 73995614
IUPAC Name tert-butyl N-[4-[4-[(Z)-N'-hydroxycarbamimidoyl]phenyl]-1,3-thiazol-2-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1=NC(=CS1)C2=CC=C(C=C2)C(=NO)N

Specifications

Melting Point 134°C to 139°C
Quantity 1 g
Formula Weight 334.39
Percent Purity 97%
Chemical Name or Material 4-[2-(Boc-amino)-4-thiazolyl]benzamidoxime
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis