missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoethyl)morpholine, 98+%

Product Code. 10733033 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10733033 5 g Glass bottle 5g
10107283 25 g Glass bottle 25g
10157460 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10733033 Supplier Thermo Scientific Acros Supplier No. 400750050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2038-03-1 | C6H14N2O | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2038-03-1
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.19
MDL Number MFCD00006182
InChI Key RWIVICVCHVMHMU-UHFFFAOYSA-N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 24°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 205°C
Flash Point 175°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4745 to 1.4765
Quantity 5 g
Specific Gravity 0.99
Synonym 2',3',4',5',6'-pentamethylacetophenone, 1-pentamethylphenyl ethanone, 1-pentamethylphenyl ethan-1-one, 2,3,4,5,6-pentamethylacetophenone, 1-2,3,4,5,6-pentamethylphenyl ethanone, acmc-1cl24, 2,3,4,5,6-pentamethyl acetophenone, 1,2,3,4,5-pentamethyl-6-acetylbenzene, 1-acetyl-2,3,4,5,6-pentamethylbenzene, ethanone,1-2,3,4,5,6-pentamethylphenyl
Solubility Information Solubility in water: soluble
SMILES NCCN1CCOCC1
IUPAC Name 2-(morpholin-4-yl)ethan-1-amine
PubChem CID 74876
Formula Weight 130.19
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material 4-(2-Aminoethyl)morpholine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-(2-Aminoethyl)morpholine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.