missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Acetoxy-ethyl)phenol, TRC

Product Code. 30138392
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138392 1 g 1g
30135955 250 mg 250mg
30004689 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138392 Supplier Toronto Research Chemicals Supplier No. TRCA1655951G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58556-55-1
Molecular Formula C10 H12 O3
Molecular Weight (g/mol) 180.2
Synonym Benzeneethanol, 4-hydroxy-, 1-acetate, Benzeneethanol, 4-hydroxy-, α-acetate (9CI), Tyrosol, acetate (6CI), 2-(4-Hydroxylphenyl)ethyl acetate, 2-(4-Hydroxyphenyl)ethanol acetate ester, 2-(4-Hydroxyphenyl)ethyl acetate, 2-(p-Hydroxyphenyl)ethyl acetate, 4-Hydroxyphenethyl acetate, Acetic acid 2-(4-hydroxyphenyl)ethyl ester, Tyrosyl acetate, 4-(2-Acetoxyethyl)phenol
IUPAC Name 2-(4-hydroxyphenyl)ethyl acetate
SMILES CC(=O)OCCc1ccc(O)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H12O3/c1-8(11)13-7-6-9-2-4-10(12)5-3-9/h2-5,12H,6-7H2,1H3
Formula Weight 180.0786
Chemical Name or Material 4-(2-Acetoxy-ethyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.