missing translation for 'onlineSavingsMsg'
Learn More

4-(2,4-Difluorobenzoyl)piperidine Hydrochloride, TRC

Product Code. 30384885
Change view
Click to view available options
Quantity:
500 mg
1 g
2 g
Unit Size:
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384885 1 g -
30387344 2 g 2g
30384064 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384885 Supplier Toronto Research Chemicals Supplier No. TRCD4457051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106266-04-0
Molecular Formula C12 H13 F2 N O . Cl H
Molecular Weight (g/mol) 261.6955
Synonym 4-(2,4-Difluorobenzoyl)piperidine hydrochloride, (2,4-Difluorophenyl)-4-piperidinylmethanone hydrochloride
IUPAC Name (2,4-difluorophenyl)-piperidin-4-ylmethanone;hydrochloride
SMILES Cl.Fc1ccc(C(=O)C2CCNCC2)c(F)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H13F2NO.ClH/c13-9-1-2-10(11(14)7-9)12(16)8-3-5-15-6-4-8;/h1-2,7-8,15H,3-6H2;1H
Formula Weight 261.0732
Chemical Name or Material 4-(2,4-Difluorobenzoyl)piperidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.