missing translation for 'onlineSavingsMsg'
Learn More

4-[(2,4-Dichloro-5-methoxyphenyl)amino]-6-methoxy-7-[3-(1-piperazinyl)propoxy]-3-quinolinecarbonitrile, TRC

Product Code. 30380952
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380952 5 mg -
30384199 50 mg -
30382388 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380952 Supplier Toronto Research Chemicals Supplier No. TRCD4353055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380843-81-2
Molecular Formula C25H27Cl2N5O3
Molecular Weight (g/mol) 516.42
Synonym N-Desmethyl Bosutinib
SMILES COc1cc(Nc2c(cnc3cc(OCCCN4CCNCC4)c(OC)cc23)C#N)c(Cl)cc1Cl

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C25H27Cl2N5O3/c1-33-22-13-21(18(26)11-19(22)27)31-25-16(14-28)15-30-20-12-24(23(34-2)10-17(20)25)35-9-3-6-32-7-4-29-5-8-32/h10-13,15,29H,3-9H2,1-2H3,(H,30,31)
Formula Weight 515.1491
Chemical Name or Material 4-[(2,4-Dichloro-5-methoxyphenyl)amino]-6-methoxy-7-[3-(1-piperazinyl)propoxy]-3-quinolinecarbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.