missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)phenyl]morpholine, 97%, Thermo Scientific™

Product Code. 10396324
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10396324 1 g Amber Glass Bottle 1g
11366805 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10396324 Supplier Thermo Scientific Maybridge Supplier No. CC57839DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-77-0
Molecular Formula C17H23BF3NO3
Molecular Weight (g/mol) 357.18
MDL Number MFCD09064984
InChI Key FGPWVOFEKZVCDA-UHFFFAOYSA-N
Synonym 4-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-4-trifluoromethyl phenyl morpholine, 4-2-tetramethyl-1,3,2-dioxaborolan-2-yl-4-trifluoromethyl phenyl morpholine, 2-morpholino-5-trifluoromethylphenylboronic acid pinacol ester, 2-morpholin-4-yl-5-trifluoromethyl benzeneboronic acid,pinacol ester
PubChem CID 24229575
IUPAC Name 4-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)phenyl]morpholine
SMILES CC1(C)OB(OC1(C)C)C1=C(C=CC(=C1)C(F)(F)F)N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 357.18
Percent Purity 97%
Chemical Name or Material 4-[2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)phenyl]morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.