missing translation for 'onlineSavingsMsg'
Learn More

4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3-pyridinecarboxylic acid Methyl Ester, TRC

Product Code. 30365575
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30365575 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30365575 Supplier Toronto Research Chemicals Supplier No. TRCD3964801MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3-pyridinecarboxylic acid Methyl Ester
CAS 253597-20-5
Molecular Formula C15 H15 Cl2 N O2
Quantity 1 mg
Synonym Methyl 4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3-pyridinecarboxylate, H 168/79
InChI Formula InChI=1S/C15H15Cl2NO2/c1-8-7-11(10-5-4-6-12(16)14(10)17)13(9(2)18-8)15(19)20-3/h4-7,11,18H,1-3H3
SMILES COC(=O)C1=C(C)NC(=CC1c2cccc(Cl)c2Cl)C
IUPAC Name methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3-carboxylate
Molecular Weight (g/mol) 312.191
Formula Weight 311.048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.