missing translation for 'onlineSavingsMsg'
Learn More

4-(2,2-Dicarboethoxy-propyl)phenylacetic Acid Ethyl Ester, TRC

Product Code. 30383671
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383671 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383671 Supplier Toronto Research Chemicals Supplier No. TRCD43099510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(2,2-Dicarboethoxy-propyl)phenylacetic Acid Ethyl Ester
CAS 223123-63-5
Molecular Formula C19 H26 O6
Quantity 10 mg
Synonym Propanedioic acid, 2-[[4-(2-ethoxy-2-oxoethyl)phenyl]methyl]-2-methyl-, 1,3-diethyl ester, Propanedioic acid, [[4-(2-ethoxy-2-oxoethyl)phenyl]methyl]methyl-, diethyl ester (9CI), 4-(2,2-Dicarboethoxypropyl)phenylacetic Acid Ethyl Ester
InChI Formula InChI=1S/C19H26O6/c1-5-23-16(20)12-14-8-10-15(11-9-14)13-19(4,17(21)24-6-2)18(22)25-7-3/h8-11H,5-7,12-13H2,1-4H3
SMILES CCOC(=O)Cc1ccc(CC(C)(C(=O)OCC)C(=O)OCC)cc1
IUPAC Name diethyl 2-[[4-(2-ethoxy-2-oxoethyl)phenyl]methyl]-2-methylpropanedioate
Molecular Weight (g/mol) 350.41
Formula Weight 350.1729
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.