missing translation for 'onlineSavingsMsg'
Learn More

4-[(1S)-3-(Dimethylamino)-1-(2-thienyl)propoxy]-1-naphthalenyl-Beta-D-glucopyranosiduronic Acid Methyl Ester, TRC

Product Code. 30386183
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386183 1 mg 1mg
30381789 5 mg 5mg
30369025 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386183 Supplier Toronto Research Chemicals Supplier No. TRCD4636151MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[(1S)-3-(Dimethylamino)-1-(2-thienyl)propoxy]-1-naphthalenyl-beta-D-glucopyranosiduronic Acid Methyl Ester
Analyte or Component Names 4-[(1S)-3-(Dimethylamino)-1-(2-thienyl)propoxy]-1-naphthalenyl-beta-D-glucopyranosiduronic Acid Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C25H29NO8S
InChI Formula InChI=1S/C25H29NO8S/c1-26-12-11-18(19-8-5-13-35-19)32-16-9-10-17(15-7-4-3-6-14(15)16)33-25-22(29)20(27)21(28)23(34-25)24(30)31-2/h3-10,13,18,20-23,25-29H,11-12H2,1-2H3/t18-,20-,21-,22+,23?,25+/m0/s1
Solution Type Neat
SMILES O[C@@H]1[C@@H](C(OC)=O)O[C@@H](OC2=C(C=CC=C3)C3=C(O[C@@H](CCNC)C4=CC=CS4)C=C2)[C@H](O)[C@H]1O
IUPAC Name methyl (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-((4-((S)-3-(methylamino)-1-(thiophen-2-yl)propoxy)naphthalen-1-yl)oxy)tetrahydro-2H-pyran-2-carboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 503.57
Quantity 1 mg
Formula Weight 503.16
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.