missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrrol-1-yl)benzonitrile, 97%, Thermo Scientific™

Product Code. p-3075519
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10481673 1 g 1g
10072494 5 g 5g
10669515 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10481673 Supplier Thermo Scientific Maybridge Supplier No. CC25516DA
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 23351-07-7
Molecular Formula C11H8N2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00085164
InChI Key OKVSZRKKRHNDOL-UHFFFAOYSA-N
Synonym 4-1h-pyrrol-1-yl benzonitrile, 1-4-cyanophenyl pyrrole, n-4-cyanophenyl pyrrole, benzonitrile, 4-pyrrol-1-yl, 4-1-pyrrolyl benzonitrile, 4-pyrrol-1-yl benzonitrile, 4-pyrrolylbenzenecarbonitrile, acmc-1cekp, maybridge1_001756, 1-4-cyanophenyl-pyrrole
PubChem CID 272424
SMILES N#CC1=CC=C(C=C1)N1C=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
IUPAC Name 4-(1H-pyrrol-1-yl)benzonitrile
Formula Weight 168.2
Percent Purity 97%
Chemical Name or Material 4-(1H-pyrrol-1-yl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.