missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrrol-1-yl)aniline, 97%, Thermo Scientific™

Product Code. 10072014
Change view
Click to view available options
Quantity:
250 mg
1 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10072014 25 g Amber Glass Bottle 25g
10252882 250 mg Amber Glass Bottle 250mg
10039401 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10072014 Supplier Thermo Scientific Maybridge Supplier No. CC25514EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 52768-17-9
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD01934575
InChI Key NHLHWHRXMZZWGA-UHFFFAOYSA-N
Synonym 4-1h-pyrrol-1-yl aniline, 4-1-pyrrolyl aniline, 4-pyrrol-1-yl aniline, benzenamine, 4-1h-pyrrol-1-yl, 4-pyrrol-1-yl-phenylamine, 4-pyrrolylphenylamine, 1-p-amino-phenylazole, 4-pyrrol-1-yl-aniline, 1-4-aminophenyl pyrrole, 1-4-amino-phenyl pyrrole
PubChem CID 2795457
IUPAC Name 4-pyrrol-1-ylaniline
SMILES C1=CN(C=C1)C2=CC=C(C=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material 4-(1H-Pyrrol-1-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.