missing translation for 'onlineSavingsMsg'
Learn More

4(1H)-Pyrimidinone, 98%, Thermo Scientific Chemicals

Product Code. 15461999
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15461999 5 g 5g
15451999 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15461999 Supplier Thermo Scientific Alfa Aesar Supplier No. H64690.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4562-27-0
Molecular Formula C4H4N2O
Molecular Weight (g/mol) 96.089
MDL Number MFCD00006664
InChI Key DNCYBUMDUBHIJZ-UHFFFAOYSA-N
Synonym pyrimidin-4-ol, 4-hydroxypyrimidine, 4-pyrimidinol, 4 3h-pyrimidone, pyrimidin-4 3h-one, 4 3h-pyrimidinone, 4-pyrimidone, 4 1h-pyrimidinone, 4-pyrimidinone, deaminoisocytosine
PubChem CID 20695
IUPAC Name 1H-pyrimidin-6-one
SMILES C1=CN=CNC1=O

Specifications

Melting Point 163°C to 169°C (literature)
Quantity 5 g
Beilstein 741972
Formula Weight 96.09
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4(1H)-Pyrimidinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.