missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-ylmethyl)benzonitrile, ≥97%, Thermo Scientific™

Product Code. 10524974
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10524974 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10524974 Supplier Thermo Scientific Maybridge Supplier No. CC56016DA
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 179057-34-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H9N3
MDL Number MFCD07687291
Quantity 1 g
Synonym 4-1h-pyrazol-1-ylmethyl benzonitrile, 4-pyrazol-1-ylmethyl benzonitrile, benzonitrile,4-1h-pyrazol-1-ylmethyl, 4-1h-pyrazol-1-yl methyl benzonitrile, 4-pyrazolylmethyl benzenecarbonitrile, 4-1h-pyrazole-1-ylmethyl benzonitrile
InChI Key FPZADNOIMZEBTC-UHFFFAOYSA-N
SMILES C1=CN(N=C1)CC2=CC=C(C=C2)C#N
IUPAC Name 4-(pyrazol-1-ylmethyl)benzonitrile
Molecular Weight (g/mol) 183.214
PubChem CID 8834695
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 4-(1H-Pyrazol-1-ylmethyl)benzonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.